missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 96-Well Semi-Skirted Microplates, BIO-CERT™ PCR QUALITY, Standard Profile

Product Code. p-4882091 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
White
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
15535980 Clear Pack of 50
15624007 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15535980 Supplier BRAND™ Supplier No. 781375
Add to basket
Edge
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

Optimized for reliable and precise results in a wide range of PCR applications.

BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Suitable for most commercially available thermal cyclers
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
    • Skirted plates provide extra rigidity, ideal for automated pipetting systems and transport
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color Clear
Disposable Single-use
Product Type PCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Semi-skirted
Material Polypropylene
Volume (Metric) Well 0.20 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) qPCR
Includes Plates only
No. per Pack 50
Profile Standard
RNase Free Yes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.