Learn More
BRAND™ Semi-Skirted Standard Profile PCR Plates
Manufactured from high-quality virgin plastics without lubricants or chemical additives that can contaminate reagents and samples. Provide excellent, reproducible results in most PCR applications.
Brand: BRAND™ 781376
162.58 GBP valid until 2024-12-31
Use promo code "21896" to get your promotional price.
Additional Details : Weight : 1.09230kg
Description
- Autoclavable
- Free of DNA, DNase, RNase, and other PCR inhibitors.
- Thin, uniform walls ensure consistent, precise thermal transfer and excellent results.
- Standard and low profile plates from BRAND are compatible with most major brands of thermal cyclers, including Applied Biosystems™ 9800 Fast, 7900 HT, and 7500 Fast, BIORAD™ CFX 96, and Eppendorf™ Mastercycler™ systems.
- White PCR plates are uniformly colored with titanium dioxide (TiO2) and have smooth surfaces to optimize reflection of the fluorescence signal.
Specifications
Standard | |
Autoclavable | |
Plastic | |
Yes | |
50 | |
Yes |
8 x 12 | |
White | |
96 | |
qPCR | |
Standard |