missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Reagent Reservoirs
Constructed of heavy-gauge polypropylene for long-lasting durability. Reagent Reservoirs are autoclavable and suitable for all 8- and 12-channel pipetters. Non-sterile version includes lid.
Brand: BRAND™ 703459
Code : 03
Additional Details : Weight : 0.23310kg
Description
- Heavy-gauge polypropylene
- Suitable for all 8- and 12-channel pipetters
- Autoclavable
- Non-sterile version includes lid
Specifications
60 mL | |
Non-sterile | |
For use with multichannel pipets. Includes lid. | |
7.11 cm | |
5.5 in. | |
Packs of 10 | |
11.68 cm |
Polypropylene | |
Reagent Reservoir | |
2.8 in. | |
Lid | |
13.97 cm | |
4.6 in. | |
Yes |