missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ 96-Well Non-Skirted Microplates, BIO-CERT™ PCR QUALITY, Standard Profile
GREENER_CHOICE

Product Code. 15311581 Shop All Brand Products
Click to view available options
Color:
Clear
White
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 15311581

Brand: BRAND™ 781369

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Optimized for reliable and precise results in a wide range of PCR applications.

BRAND™ PCR products deliver excellent, reproducible results for a wide range of PCR and qPCR applications. Manufactured from high-quality virgin resins, these products are free from lubricants, chemical additives, DNA, DNase, RNase, and other PCR inhibitors. Key features include:

  • Exceptional Quality and Purity
    • BIO-CERT™ PCR QUALITY ensures plates are free of human DNA, RNase, DNase, pyrogens, and PCR inhibitors
    • Free of contaminants that could interfere with reagents and samples
    • Ultra-smooth vessel interiors minimize enzyme nucleic acid binding
  • Precision Thermal Performance
    • Thin, uniform walls ensure consistent, precise thermal transfer for reliable results
    • Designed for rapid and accurate heat transfer, reducing cycle times
  • Wide Compatibility
    • Non-skirted plates are highly flexible and compatible with most thermal cyclers
    • White PCR plates, enhanced with titanium dioxide (TiO2), optimize fluorescence signal reflection and are recommended for qPCR applications
  • User-Friendly Features
    • Blue alphanumeric coding for easy sample identification
    • Well rims designed to prevent cross-contamination and ensure reliable sealing with tailored mats
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Untreated
Certifications/Compliance DNA, DNase, RNase-free
Format Standard
Array 8 x 12
Autoclavable Autoclavable
Packaging Bagged
Color White
Disposable Single-use
Product Type qPCR 96-well plate
For Use With (Equipment) Thermal Cyclers
Skirt Style Non-skirted
Material Polypropylene
Volume (Metric) Well 0.20 mL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Non-treated
Well Shape Conical Bottom (V-well)
Dnase Free Yes
For Use With (Application) qPCR
Includes Plates only
No. per Case 50
No. per Pack 50/Pk.
Profile Standard
RNase Free Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.