missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Non-Sterile Microcentrifuge Tubes

Product Code. 10232481 Shop All Brand Products
Change view
Click to view available options
Color:
Amber
Blue
Clear
Green
Orange
Yellow
Quantity:
2000/Pk.
500/Pk.
Unit Size:
Pack of 2000
Pack of 500
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Quantity unitSize
10232481 Yellow 500/Pk. Pack of 500
10478502 Clear 500/Pk. Pack of 500
11778218 Clear 2000/Pk. Pack of 2000
10749341 Blue 500/Pk. Pack of 500
10243051 Green 500/Pk. Pack of 500
10468892 Orange 500/Pk. Pack of 500
10478892 Amber 500/Pk. Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 10232481 Supplier BRAND™ Supplier No. 780521

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Fits most common rotors and may be spun up to 30,000 RCF. Non-Sterile Microcentrifuge Tubes are highly translucent and free from flash, air bubbles, and streaks. Polypropylene flat snap caps form a tight, leak-free, easy-open seal. Tubes have a uniform wall thickness for consistent, repeatable results.

Fits most common rotors and may be spun up to 30,000 RCF (g) (10,000 x g for 0.5mL; 20,000 x g for 1.5mL; 30,000 x g for 1.5mL and 2.0mL with lid closure at 20°C for 20 min).
  • Manufactured from virgin polypropylene
  • Highly translucent tubes are free from flash, air bubbles, and streaks.
  • Caps form a tight, leak-free seal that is easy to open, even when wearing gloves, and are flat with consistent thickness for trouble-free piercing.
  • Tubes have uniform wall thickness for consistent, repeatable results.
  • Subdivisions for approximate volume determination.
  • Autoclavable at 120°C (2bar), according to DIN EN 285.
  • Lid closure are certified endotoxin, DNA-, DNase-, and RNase-free.
  • Colored tubes simplify sample identification
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Endotoxin, RNase-, ATP- and DNA-free
Color Yellow
Volume (Metric) 1.5 mL
Quantity 500/Pk.
Closure Material Polypropylene
For Use With (Equipment) Centrifuge
Sterility Non-sterile
Material Polypropylene
Max. Force 20,000 RCF
Closure Type Flat Snap Cap
Autoclavable Autoclavable
Format Tube
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.