missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates immunoGrade™ 96-Well Microplates

Product Code. 10173853 Shop All Brand Products
Change view
Click to view available options
Color:
Black
Black, Clear Bottom
Clear
White
Well Shape:
Conical Bottom (V-well)
Flat Bottom (F-well)
Round
Unit Size:
Pack of 100
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape unitSize
10173853 Black, Clear Bottom Flat Bottom (F-well) Pack of 100
10193563 Clear Round Pack of 100
10647143 Clear Conical Bottom (V-well) Pack of 100
10032113 Clear Flat Bottom (F-well) Pack of 100
10477863 White Round Pack of 100
10438623 Black Round Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 10173853 Supplier BRAND™ Supplier No. 781731
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

An excellent choice for the majority of standard ELISAs as well as solid phase immunoassays

  • Optimized for the immobilization of IgG
  • High binding capacity for molecules with mixed hydrophobic and hydrophilic regions
  • Designed to minimize cross-contamination
  • Blue alphanumeric embossing for easy well identification on standard plates
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Black, Clear Bottom
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment immunoGrade
Total Volume (Metric) Well 330 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) For the majority of standard ELISAs as well as solid phase immunoassays
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.