missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ premium, 96-Well Flat-Bottom Microplates, PS

Product Code. 10032403 Shop All Brand Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10032403 White Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10032403 Supplier BRAND™ Supplier No. 782090
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

cellGrade™ premium surface treatment is ideal for cultivating demanding cell lines in serum-free or serum-reduced environments. Provides a reliable alternative to biologically coated surfaces with good room temperature shelf life.

  • Poly-D-Lysine-equivalent surface delivers similar cell growth and morphology
  • Optimized cell adhesion reduces damage from frequent washing
  • Suitable for for serum-free and serum-reduced cultivation
  • Physical/chemical treatment offers a longer shelf life at room temperature compared to biological coatings
  • Sterilized by B-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Complies with Ph. Eur. and USP 29 standards
  • BIO-CERT™ CELL CULTURE QUALITY: DNA-, DNase-, RNase-free and non-cytoxic
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

Order Info

Packs of 50 individually wrapped plates with lids

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ISO 11137, USP 29
Format Standard
Lid With Lid
Array 8 x 12
Color White
Product Type 96-well plate
Material Polystyrene
Volume (Metric) Well 330 μL
No. of Wells 96
Sterility Sterile
Surface Treatment cellGrade Premium
Total Volume (Metric) Well 330 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cell lines with the highest demands on their environment
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.