missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 384-Well, Flat-Bottom Microplate

Product Code. 10781924 Shop All Brand Products
Click to view available options
Color:
Black
Unit Size:
Pack of 50
This item is not returnable. View return policy

Product Code. 10781924

Brand: BRAND™ 781987

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Black; Sterility:Sterile; Lid:With Lid

  • Polystyrene construction
  • Sterile
  • Lids included
  • Standard plate for the cultivation of adherent cell lines
  • PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible
  • Surface is hydrophilic compared with non-treated PS
  • Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells
  • Clearly distinguishable through the color code: orange, embossed alphanumeric coding for 96-well standard plates
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SLAS
Format Standard
Lid With Lid
Array 16 x 24
Binding Type Carboxyl and Hydroxyl Groups
Color Black
Material Polystyrene
No. of Wells 384
Sterility Sterile
Total Volume (Metric) Well 120 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) Cultivation of adherent cell lines
No. per Pack 50
RNase Free Yes
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.