missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 384-Well Flat-Bottom Microplates, PS
Shop All Brand Products
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
Description
- Polystyrene construction
- Sterile with included lids
- Designed for the cultivation of adherent cell lines
- Hydrophilic PS surface with accessible carboxyl and hydroxyl groups
- Enhance serum component binding for indirect cell adhesion
- Clearly distinguishable with orange color code and embossed alphanumeric coding for 384-well plates
- BIO-CERT™ CELL CULTURE QUALITY: free from endotoxins, DNase, DNA, RNase, non-cytotoxic
Specifications
Specifications
| Barcode | No Barcode |
| Certifications/Compliance | ANSI, SLAS |
| Format | Standard |
| Lid | With Lid |
| Array | 16 x 24 |
| Autoclavable | Not Autoclavable |
| Binding Type | Carboxyl and Hydroxyl Groups |
| Product Line | BRANDplates |
| Color | Clear |
| Product Type | 384-well microplate |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction