missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 1536-Well, cellGrade-Treated, Flat-Bottom Microplate

Product Code. 10194283 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
10194283 Clear Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10194283 Supplier BRAND™ Supplier No. 782000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Color:Clear; Sterility:Sterile; Lid:With Lid

  • Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
  • Easily binds serum components to for indirect adhesion of cells
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet the requirements of Ph. Eur. and the USP 29
  • DNA-, DNase-, RNase-free; noncytotoxic
  • Endotoxins are <0.01 EU/mL
  • For standard cell culture applications
  • Individually wrapped with lid

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SLAS
Format Standard
Lid With Lid
Array 32 x 48
Binding Type Carboxyl and Hydroxyl Groups
Color Clear
Material Polystyrene
No. of Wells 1536
Sterility Sterile
Surface Treatment cellGrade
Total Volume (Metric) Well 10 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of adherent cell lines
No. per Case 50/Cs.
No. per Pack 1/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.