missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ BRANDplates cellGrade™ 1536-Well, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Brand: BRAND™ 782002
Code : 03
Additional Details : Weight : 2.22000kg
Description
- Hydrophilic surface with freely-accessible carboxyl and hydroxyl groups
- Easily binds serum components to for indirect adhesion of cells
- Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
- Meet the requirements of Ph. Eur. and the USP 29
- DNA-, DNase-, RNase-free; noncytotoxic
- Endotoxins are <0.01 EU/mL
- For standard cell culture applications
- Individually wrapped with lid
Specifications
ANSI, SLAS | |
With Lid | |
Carboxyl and Hydroxyl Groups | |
Polystyrene | |
Sterile | |
Flat Bottom (F-well) |
Standard | |
32 x 48 | |
Black | |
1536 | |
10 μL | |
Cultivation of adherent cell lines |