Learn More
BRAND™ BRANDplates™ 384-Well Microplate
Product Code. 15464163 Shop All Brand Products
Change view
Click to view available options
Color:
White
Unit Size:
Pack of 5
Product Code. Color unitSize
15464163 White Pack of 5
1 options
Product Code. 15464163 Supplier BRAND™ Supplier No. 781988
Only null left
Add to basket
Add to basket
BRANDplates™ 384-Well Microplate with cellGrade™ surface treatment and F-bottom.
  • Standard plate for the cultivation of adherent cell lines
  • PS-surface with different chemical groups, such as carboxyl and hydroxyl groups, that are freely accessible
  • Surface is hydrophilic compared with non-treated PS
  • Serum components are easily bound onto the freely accessible chemical groups, allowing an indirect adhesion of cells
  • Free from endotoxins, DNase, DNA, RNase, non-cytotoxic
  • Sterile
  • With lids
TRUSTED_SUSTAINABILITY
Specifications
Lid With Lid
Color White
Material Polystyrene
No. of Wells 384
Sterility Sterile
Surface Treatment cellGrade
Total Volume (Metric) Well 120 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
RNase Free Yes
Well Color Transparent