missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates™ Smooth-Walled Insert Strips

Product Code. 15690637 Shop All Brand Products
Click to view available options
Pore Size:
0.4 μm
1 μm
3 μm
8 μm
Supportive Membrane:
PC
PET
Unit Size:
Pack of 12
This item is not returnable. View return policy

Product Code. 15690637

Brand: BRAND™ 782870

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Available with either polycarbonate (PC) or polyethylene terephthalate (PET) membranes.

  • Sterile
  • BIO-CERT™ CELL CULTURE QUALITY: Free of human DNA, RNase, DNase, pyrogens, and non-cytotoxic
  • Smooth-walled
  • cellGrade™ plus surface treatment
  • Available in PC or PET membrane materials for different optical requirements and cell visibility
  • Multiple membrane pore sizes for different applications
  • Packaged in 12 strips of four wells per pack

Applications:

Secretion studies, co-culture, migration studies, chemotaxis assays

TRUSTED_SUSTAINABILITY

Specifications

Bottom Shape Flat
Capacity (Metric) 400 μL
Color Clear
Culture Area 0.6 cm2
Material Polystyrene
Sterility Sterile
Surface Treatment cellGrade plus
Type Cell Culture Insert Strip
Well Count 4
Pore Size 8 μm
Quantity 12/Pk.
Supportive Membrane PET
Height (Metric) 13.4 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.