missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates lipoGrade™ 96-Well Microplates

Product Code. p-3816208 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape unitSize
10772884 Clear Flat Bottom (F-well) Pack of 100
10752884 Clear Round Bottom (U-well) Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10772884 Supplier BRAND™ Supplier No. 781842
Add to basket
Add to basket
This item is not returnable. View return policy

Immunoassay plate optimized for the immobilization of hydrophobic molecules

  • High binding for molecules with predominately hydrophobic regions such as lipoproteins or peptides
  • PS, non-sterile
  • Blue alphanumeric embossing on clear, white and black solid plates for easy well identification
  • For solid phase assays with hydrophobic molecules and liquid phase with hydrophilic molecules, respectively
  • Alternative to immunoGrade™ surface
TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Binding Type High Binding
Color Clear
Product Type 96-well Microtitration Plate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment lipoGrade
Well Shape Flat Bottom (F-well)
For Use With (Application) Solid phase with hydrophobic molecules and liquid phase with hydrophilic molecules
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.