missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates hydroGrade™ 96-Well Microplates

Product Code. 10206813 Shop All Brand Products
Change view
Click to view available options
Color:
Clear
Well Shape:
Flat Bottom (F-well)
Round Bottom (U-well)
Total Volume (Metric) Well:
330 μL
350 μL
Unit Size:
Pack of 100
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Well Shape Total Volume (Metric) Well unitSize
10206813 Clear Flat Bottom (F-well) 350 μL Pack of 100
10012403 Clear Round Bottom (U-well) 330 μL Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10206813 Supplier BRAND™ Supplier No. 781782
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Featuring a strongly hydrophilic surface for binding of hydrophilic molecules or for homogeneous assays with hydrophobic molecules that remain in solution

  • Immunoassay surface with strong affinity to hydrophilic molecules, such as glycoproteins and peptides, antibodies with predominantly hydrophilic regions, and nucleic acids
  • Blue alphanumeric embossing for easy well identification
  • Non-sterile

Order Info

Packaging is 20 stacks of 5 plates each (100 plates). Each stack comes with one lid.

TRUSTED_SUSTAINABILITY

Specifications

Barcode No Barcode
Binding Property Hydrophilic
Certifications/Compliance ANSI, SLAS
Format Standard
Lid Without Lid
Array 8 x 12
Color Clear
Product Type 96-well Microplate
Material Polystyrene
Volume (Metric) Well 350 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment hydroGrade
Total Volume (Metric) Well 350 μL
Well Shape Flat Bottom (F-well)
For Use With (Application) Immobilization of hydrophilic molecules
Quantity 100/Cs.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.