missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ BRANDplates cellGrade™ 96-Well, Cell Culture-Treated, Flat-Bottom Microplate

Product Code. 15624027 Shop All Brand Products
Click to view available options
Color:
Black, Clear Bottom
Unit Size:
Pack of 5
This item is not returnable. View return policy

Product Code. 15624027

Brand: BRAND™ 782095

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Color:Black, Clear Bottom; Sterility:Sterile; Lid:With Lid

  • Poly-D-Lysine-equivalent surface with analogous results regarding cell growth and morphology
  • Optimized cell adhesion reduces cell damage from frequent washing
  • Allows for serum-free and serum-reduced cultivation
  • Physical/chemical treatment has long room temperature shelf life compared to biological coatings
  • Sterilized by β-radiation to ISO 11137 and AAMI guidelines for a SAL of 10-6
  • Meet requirements of Ph. Eur. and USP 29
  • DNA-, DNase-, RNase-free
  • Endotoxins are <0.01 EU/mL
  • Orange alphanumeric embossing for easy well identification

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ISO 11137, USP 29
Lid With Lid
Color Black, Clear Bottom
Material Polystyrene
No. of Wells 96
Sterility Sterile
Surface Treatment Cell Culture
Well Shape Flat Bottom (F-well)
For Use With (Application) Cultivation of cell lines with the highest demands on their environment
No. per Pack 5
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.