missing translation for 'onlineSavingsMsg'
Learn More

BRAND™ Sterile Ultra Low Retention Polypropylene Pipet Tips

Product Code. 15144937 Shop All Brand Products
Change view
Click to view available options
Format:
TipBox
TipStack
Unit Size:
Pack of 960
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Format Unit Size
15144937 TipBox Pack of 960
15134947 TipStack Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15144937 Supplier BRAND™ Supplier No. 732328
Quantity Price
1 £79.87 / Pack of 960
10 £75.32 / Pack of 960

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Significantly reduces sample loss and provides substantially higher reproducibility when working with critical media.

  • Ideal for biological samples that contain detergents such as TRITON™ X-100
  • No additives that can be leached out and no siliconization of the surface
  • High chemical resistance, ideal for working with solvents
  • Tips can be autoclaved at 121°C without damaging the material properties

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 2 to 200 μL
Sterility Sterile
Color Colorless
Autoclavable Autoclavable
Certifications/Compliance DNA, RNase, ATP and endotoxin-free
Clean Claims DNase, RNase, ATP, and Endotoxin Free
For Use With (Application) High translucency, easy ejection and low waste, quick volume check
Format TipBox
Tip Style Ultra Low Retention
Graduated Yes
Material Polypropylene
Graduations 20, 100μL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.