missing translation for 'onlineSavingsMsg'
Learn More

Branaplam, MedChemExpress

Product Code. 16443777
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16443777

Brand: Medchem Express HY19620

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Branaplam (LMI070; NVS-SM1) is a highly potent, selective and orally active survival motor neuron-2 (SMN2) splicing modulator with an EC50 of 20 nM for SMN. Branaplam inhibits human-ether-a-go-go-related gene (hERG) with an IC50 of 6.3 μM. Branaplam elevates full-length SMN protein and extends survival in a severe spinal muscular atrophy (SMA) mouse model.

Branaplam (LMI070

Specifications

Chemical Name or Material Branaplam
CAS 1562338-42-4
Color Yellow
Purity Grade Notes Research
Molecular Formula C22H27N5O2
Quantity 50 mg
Synonym LMI070 NVS-SM1
Solubility Information DMSO : 5 mg/mL (12.71 mM; ultrasonic and warming and heat to 80°C)
SMILES OC1=CC(C2=CNN=C2)=CC=C1C3=CC=C(OC4CC(C)(C)NC(C)(C)C4)N=N3
Molecular Weight (g/mol) 393.48
Formula Weight 393.48
Percent Purity 95.62%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.