missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ Brain Temporal Lobe Tissue Slides (Parkinson's)- Paraffin

Product Code. 18605211 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
Product Code. Quantity unitSize
18605211 5 Slides Pack of 5
1 options
This item is not returnable. View return policy

Product Code. 18605211

Brand: Novus Biologicals™ NBP278008

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tissue Slides

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Slides
For Use With (Application) Immunohistochemistry (IHC), Immunohistochemistry (Paraffin), ISH, Dual RNAScope ISH-IHC
Species Human
Target Specificity Brain Temporal Lobe
Content And Storage Store at 4°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.