missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Brain Medulla Oblongata Tissue Slides (Multiple Sclerosis)- Frozen

Product Code. 18620371 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 Slides
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18620371 5 Slides Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18620371 Supplier R&D Systems™ Supplier No. NBP277681
  • ONLINE EXCLUSIVE PRICE
    £717.69
    £615.30 / Pack of 5
    Save £102.39
    14% Off
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Tissue Slides

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 Slides
For Use With (Application) Immunohistochemistry (IHC), Immunohistochemistry (Frozen), ISH
Product Line Novus
Species Human
Target Specificity Brain Medulla Oblongata
Content And Storage Store at -70°C. Avoid freeze-thaw cycles.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.