missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BRAF35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5148289
This item is not returnable.
View return policy
Description
BRAF35 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
Required for correct progression through G2 phase of the cell cycle and entry into mitosis. Required for RCOR1/CoREST mediated repression of neuronal specific gene promoters.
Specifications
| BRAF35 | |
| Polyclonal | |
| Unconjugated | |
| BRAF | |
| AW610687; BRAF25; BRAF35; BRCA2-associated factor 35; fc85b02; FLJ26127; high mobility group 20 B; high mobility group 20B; high mobility group box domain containing; high mobility group protein 20 B; high-mobility group 20B; HMG box domain containing 2; HMG box-containing protein 20B; HMG domain-containing protein 2; HMG domain-containing protein HMGX2; HMG20B; Hmgx2; HMGXB2; hypothetical protein LOC553572; PP7706; pp8857; si:dkey-110c1.6; SMARCE1R; SMARCE1-related protein; SOXL; Sox-like transcriptional factor; Structural DNA-binding protein BRAF35; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; LOW QUALITY PROTEIN: SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E member 1-related; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily E, member 1-related; Unknown (protein for MGC:134468); wu:fc85b02; zgc:110001 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 10362, 673 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 100 μg/mL | |
| PBS with 0.03% ProClin 300; pH 7 | |
| P15056, Q9P0W2 | |
| BRAF, HMG20B | |
| E. coli-derived Human HMG20B fragment. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction