missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAF35 Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB200-126FR
This item is not returnable.
View return policy
Description
BRAF35 Polyclonal antibody specifically detects BRAF35 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| BRAF35 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| DyLight 680 | |
| BRAF35Sox-like transcriptional factor, high-mobility group 20B, HMG domain-containing protein 2, HMG domain-containing protein HMGX2, HMGX2FLJ26127, HMGXB2member 1-related, SMARCE1R, SMARCE1-related protein | |
| Residues 31-50 [KQERGEGPRA GEKGSHEEEP] of the human structural DNA-binding protein BRAF35. | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Cycle and Replication, Tumor Suppressors | |
| 10362 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction