missing translation for 'onlineSavingsMsg'
Learn More

BRAF Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech

Product Code. 18095495 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18095495 20 μL 20µL
18090726 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18095495 Supplier Proteintech Supplier No. CL4882089920UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The BRAF antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human BRAF. This antibody recognizes human, mouse antigen. The BRAF antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.

Specifications

Antigen BRAF
Applications Immunofluorescence
Concentration 1000 μg/mL
Conjugate CoraLite Plus 488
Gene Symbols B RAF1, BRAF, BRAF1, FLJ95109, p94, Proto oncogene B Raf, RAFB1
Host Species Rabbit
Quantity 20 μL
Primary or Secondary Primary
Gene ID (Entrez) 673
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.