missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bradykinin RB1/BDKRB1 Antibody, CoraFluor™ 1, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-99565CL1
This item is not returnable.
View return policy
Description
Bradykinin RB1/BDKRB1 Polyclonal antibody specifically detects Bradykinin RB1/BDKRB1 in Human samples. It is validated for Western Blot
Specifications
| Bradykinin RB1/BDKRB1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| CoraFluor 1 | |
| B1 bradykinin receptor, B1BKR, B1R, BK-1 receptor, BKB1R, BKR1, bradyb1, bradykinin B1 receptor, bradykinin receptor 1, bradykinin receptor B1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human Bradykinin RB1/BDKRB1. | |
| 0.1 mL | |
| GPCR, Immunology, Innate Immunity, Neuroscience | |
| 623 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction