missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyJet J4000 Color Label Printer with Safety and Facility ID Software

Product Code. 17459242
Click to view available options
Color:
Blue, Gray
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 17459242

Brand: Brady™ 310442

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This industry inkjet workhorse creates up to 5,000 impactful full-color labels per day up to 203.20 mm wide on continuous or die-cut material.

  • This industry inkjet workhorse creates up to 5,000 impactful full-colour labels per day up to 203.20 mm wide on continuous or die-cut material
  • Ideal for a wide variety of applications in safety, including Lean / 5S, lockout tagout, instructional or safety chemical labelling for handling, storage or transport, as well as lab settings
  • Contains the color options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels
  • Streamline storage by only needing to stock white label material and print only what you need, when you need it, in photo-quality full colour every time
  • Easily print PDFs or from third-party software packages with the optimised Windows driver
  • Includes Brady Workstation Safety and Facility Identification Software suite, enabling you to immediately start designing professional signs and labels

Specifications

Product Type Printer
Color Blue, Gray
Display Type LED
For Use With (Equipment) Brady Labels
Material Plastic
Print Technology Inkjet
Length (English) 9.1 in.
Width (English) 17.25 in.
Length (Metric) 231.14 mm
Width (Metric) 438.15 mm
Depth (English) 17.25 in.
Depth (Metric) 438.15 mm
Color Capability Full Color
Description US Plug Type
Includes 1 x BradyJet J4000 Printer, 1 x Power Cord, 1 x CMY Ink Cartridge with built-in Printhead, 1 x Pack of 2 Ink Cartridge storage caps, 1 x Media starter roll 101.60 mm wide B-2595 Vinyl, 1 x USB cord, 1 x USB Flash Drive with product information, 1 x Brady WorkStation SFID Suite
Model J4000
Print Speed 114.30 mm/s
Resolution 4800 dpi
Connector Types USB, Ethernet
Weight (English) 36 lb.
Weight (Metric) 16 kg
Certifications/Compliance CE, C-UL US, FCC Class A, RoHS directive 2011/65/EU, UL Listed for US
For Use With (Application) Facility identification, General identification, Hazard communication identification, Lean and 5S labelling, Pipe and valve marking, Warehouse marking
Quantity Each
Warranty 1 Year
Power Supply Details AC Adapter
Voltage 100/240 V
Keyboard Type None
Alternate Part Number J4000-US-BWSSFID
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.