missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyJet J4000 Color Label Printer UK

Product Code. 17469212
Change view
Click to view available options
Print Technology:
Inkjet
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Print Technology unitSize
17469212 Inkjet Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17469212 Supplier Brady™ Supplier No. 152712
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Create a safer, more efficient workplace with compliant, photo quality, full-color labels, signs and procedures that last.

This industry inkjet workhorse creates up to 5000 impactful, full-color labels per day up to 203.2 mm wide on continuous or die-cut material.

  • Ideal for a wide variety of applications in safety, including lockout tagout, instructional or safety chemical labelling for handling, storage or transport, and lab settings
  • Contains the color options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels
  • Streamline storage by only needing to stock white label material and printing only what you need, when you need it, in photo quality full color every time
  • Easily print PDFs or from third-party software packages with the optimized Windows™ driver

Specifications

Display Type LED
Print Technology Inkjet
Width (English) 17.2 in.
Width (Metric) 438 mm
Height (English) 9 in.
Height (Metric) 231 mm
Depth (English) 17.2 in.
Depth (Metric) 438 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Multi-color
Includes BradyJet J4000 Printer, UK Power Cord, CMY Ink Cartridge with Printhead, 2 Ink Cartridge Storage Caps, Media Starter Roll, USB Cord, USB Flash Drive with Product Information, Brady™ Workstation Basic Suite
Model J4000
Print Speed 114.3 mm/s
Resolution 4800 dpi
Connector Types Ethernet, USB
Operating System Windows™ 7, Windows™ 8, Windows™ 10
Temperature Range (Metric) 15°C to 32°C
Weight (English) 35 lb.
Weight (Metric) 16 kg
For Use With (Application) Facility Identification, General Identification, Hazard Identification, Warehouse Marking
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number J4000-UK
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.