missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BradyJet J4000 Color Label Printer Kit UK with Workstation SFID Suite

Product Code. 17429242
Change view
Click to view available options
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
17429242 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17429242 Supplier Brady™ Supplier No. 310391

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Create a safer, more efficient workplace with compliant, photo quality, full-color labels, signs and procedures that last.

This industry inkjet workhorse creates up to 5000 impactful, full-color labels per day up to 203.2 mm wide on continuous or die-cut material.

  • Ideal for a wide variety of applications in safety, including lockout tagout, instructional or safety chemical labelling for handling, storage or transport, and lab settings
  • Contains the color options you need to create ANSI, GHS, ISO, BS5609 and arc flash compliant labels
  • Streamline storage by only needing to stock white label material and printing only what you need, when you need it, in photo quality full color every time
  • Easily print PDFs or from third-party software packages with the optimized Windows™ driver
  • Includes UK power cord and Brady™ Workstation Safety & Facility Identification Suite

Specifications

Display Type LED
Print Technology Inkjet
Width (English) 17 in.
Width (Metric) 432 mm
Height (English) 9 in.
Height (Metric) 229 mm
Depth (English) 17 in.
Depth (Metric) 432 mm
Autoclavable Not Autoclavable
Sterility Non-sterile
Color Capability Multi-color
Includes BradyJet J4000 Printer, UK Power Cord, CMY Ink Cartridge with Printhead, 2 Ink Cartridge Storage Caps, Media Starter Roll, USB Cord, USB Flash Drive with Product Information, Brady™ Workstation SFID Suite
Model J4000
Print Speed 114.3 mm/s
Resolution 4800 dpi
Connector Types Ethernet, USB
Operating System Windows™ 7, Windows™ 8, Windows™ 10
Temperature Range (Metric) 15°C to 32°C
Weight (English) 35 lb.
Weight (Metric) 16 kg
For Use With (Application) Facility Identification, General Identification, Hazard Identification, Warehouse Marking
Warranty 1 Year
Voltage 110/240 V
Alternate Part Number J4000-UK-BWSSFID
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.