missing translation for 'onlineSavingsMsg'
Learn More

Brady™ ToughStripe™ Anti-Slip Floor Safety Sign

Product Code. 11721715
Change view
Click to view available options
Displayed Text:
BIOHAZARD
Chemical Storage
Ear Protection Area
Emergency Exit
Exit
Eye Protection Area
Eye Wash Station
Fire Alarm
Fire Extinguisher
Fire Hose
First Aid
Hard Hat Area
Safety Glasses Required
Safety Goggles Required
Safety Shower
Watch Your Step
Color:
Black
Black on Orange
Black on White
Black on Yellow
Red
White
Shape:
Rectangular
Round
Unit Size:
Each
17 product options available for selection
Product selection table with 17 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Displayed Text Color Shape unitSize
11721715 Watch Your Step Black on Yellow Rectangular Each
11701705 Safety Shower Black Round Each
11791695 First Aid White Round Each
11731705 Exit Black on White Round Each
11741705 First Aid White Rectangular Each
11771705 Fire Hose Black on White Round Each
11751705 Fire Extinguisher Black on White Round Each
11781705 Emergency Exit Black on White Round Each
11761705 Fire Alarm Black on White Round Each
11711715 Chemical Storage Red Rectangular Each
11756355 Ear Protection Area Black on Yellow Rectangular Each
11766355 Eye Protection Area Black on Yellow Rectangular Each
11776355 Hard Hat Area Black on Yellow Rectangular Each
11796355 BIOHAZARD Black on Orange Rectangular Each
11786355 Safety Goggles Required Black on Yellow Rectangular Each
11706365 Eye Wash Station Black Round Each
11736365 Safety Glasses Required Black on Yellow Rectangular Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 17 options available.
17 options
This item is not returnable. View return policy
Product Code. 11721715 Supplier Brady™ Supplier No. 104499

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Hard-to-miss signs made of durable floor marking tape

  • Backed with high-performance adhesive
  • Features a highly visible, glossy appearance for communicating important safety and warning messages to employees
  • Clear anti-skid area
  • Designed with sections of anti-skid material that help prevent accidental slips, trips and falls
  • Easy to remove and apply
  • Made of polyester

Specifications

Certifications/Compliance ASTM ANSI F1679, ANSI A1264.2-2006, OSHA
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Permanent Rubber
Description ToughStripe - Pre-printed Floor Signs
For Use With (Application) Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements with glossy appearance and clear anti-skid area for communicating important safety and warning messages to employees
Height (English) 14 in.
Height (Metric) 355.60 mm
Max. Temperature (English) 129.9°F
Min. Temperature (English) -0.4°F
Thickness 0.01 in.
Width (English) 18 in.
Width (Metric) 457.20 mm
Temperature Range (Metric) -18°C to 54°C
Shape Rectangular
Temperature Range (English) -0.4°F to 129.9 °F
Languages English
Dimensions (L x W) 457.20 x 355.60 mm
Displayed Text Watch Your Step
Product Type Floor Marking Safety Sign
Color Black on Yellow
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.