Learn More
Brady™ TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels
TLS2200™ and TLS-PC™ Vial, Centrifuge or Eppendorf Tubes™ Labels are ideal for cables, laboratory ID or general labeling where graphics need to be protected.
Brand:  Brady™ 062259
Additional Details : Weight : 0.04310kg
 View more versions of this product
Product Code. 11532905
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Description
Description
  • Labels for the TLS 2200™ and TLS-PC Link™ thermal transfer Portable Printers.
  • RoHS compliant to 2005/618/EC MCV amendment to RoHS Directive 2002/95/EC.
  • Matte white polyamide coated nylon cloth material (B-499) has a permanent acrylic adhesive with a service temperature of -40 to 193° F (-40 to 90° C) and recommended for indoor use.
  • Self-Laminating Polyester (B-461) has a permanent acrylic adhesive , good print smudge resistance, solvent resistance and high/low temperature performance.
Specifications
Specifications
RoHS
Acrylic, Permanent
Matte
Vinyl
Centrifuge tube, Cryo vial, Eppendorf tube, Laboratory identification, PCR tube
100/Roll
-40 to 158°F
0.004 in.
60.33 mm
2.2 in.
0.5 in.
TLS 2200/TLS PC Link Labels
White
Label
TLS 2200™/TLS-PC Link™ Label
1
Rectangle
-40°C to +70°C
0.102 mm
3 to 000 AWG
12.70 mm
55.88 mm