missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Signs: Production Area

Product Code. 11766365
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11766365

Brand: Brady™ 104513

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

PRODUCTION AREA ToughStripe™ Floor Signs feature a highly visible, glossy appearance for communicating important safety messages and anti-skid sections to help prevent slips, trips and falls.

  • ToughStripe™ Floor Signs communicate important safety and warning messages to your employees in areas with foot traffic
  • Durable ToughStripe™ Anti-Skid Polyester (B-534) includes a rigid, clear overlaminate and is selectively covered by anti-skid tread
  • Highly visible, glossy material resists harsh chemicals and is easy to apply and remove
  • Floor signs alert workers of potential hazards, direct traffic, and enforce protective equipment requirements.

Specifications

Certifications/Compliance ANSI Z53.1-1967; ANSI Z535-2006; OSHA 1910.145
Finish Gloss
Material Polyester
Quantity 1 Ea.
Adhesive Type Rubber-based
Description ToughStripe - Pre-printed Floor Signs
Diameter (English) 17 in.
Diameter (Metric) 431.80 mm
For Use With (Application) Used to mark aisles, passageways, storage locations, point out safety equipment and exits and enforce protective equipment requirements
Max. Temperature (English) 129.9°F
Min. Temperature (English) -0.4°F
Thickness 0.01 in.
Temperature Range (Metric) -18°C to 54°C
Temperature Range (English) -0.4°F to 129.9 °F
Languages English
Displayed Text Production Area All Through Traffic Use Designated Walk Way
Product Type Floor Safety Sign
Color Black on Yellow
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.