Learn More
Brady™ LABXPERT™ Polyester Slide Labels
Use these labels for laboratory and healthcare applications. Brady™ LABXPERT™ Polyester Slide Labels are for use with LABXPERT™ and IDXPERT™ printers.
£54.90
Specifications
Certifications/Compliance AGA, UL, UL969, CSA, C22.2 No.0.15-95, RoHS, 2005/618/EC MCV, 2002/95/EC
Description Surface Mount
Adhesive Type Acrylic, Permanent
Color Black, White
Finish Matte
View More Specs
Products 1
Product Code Brand Height (Metric) Width (Metric) Price Quantity & Availability  
Product Code Brand Height (Metric) Width (Metric) Price Quantity & Availability  
10123092
missing translation for 'viewDocuments' Promotion Details
Brady™
060952
6.35 mm 4.82 cm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
missing translation for 'toSaveUpto' 
»
Description
Description
  • Withstand temperatures from -80°C to +121°C (-112°F to +250°F)
  • Material offers permanent adhesion, outstanding chemical resistance, and write-on capabilities
  • Resist xylene, toluene, DMSO, and ethanol
  • Specifications
    Specifications
    AGA, UL, UL969, CSA, C22.2 No.0.15-95, RoHS, 2005/618/EC MCV, 2002/95/EC
    Acrylic, Permanent
    Matte
    Polyester
    Asset tracking, Barcoding, Component ID, Equipment ID, General purpose ID, Rating plates, Slide label
    Surface Mount
    Black, White
    Label
    Label
    iDXPERT™, LabXpert™ Printer