missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ “Danger: Acetylene” Signs
“Danger: Acetylene” Signs from Brady™ are ideal for use in light duty industrial, utility, commercial and institutional environments.
£112.00
Specifications
Certifications/Compliance | ANSI Z535, OSHA 1910.145 |
---|---|
Quantity | 1 Ea. |
Mounting Type | Surface Mount |
Min. Temperature (English) | -40°F |
Displayed Text | DANGER Acetylene |
Product Code | Brand | Material | For Use With (Application) | Thickness | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Material | For Use With (Application) | Thickness | Price | Quantity & Availability | |||
15339950
|
Brady™
7020 6 |
Fiberglass | Industrial, utility, commercial and institutional environment | 0.1 in. |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
- Self-Sticking Polyester (B-302) is an Indoor/Outdoor high performance pressure sensitive sign that is surface printed polyester with a polyester overlaminate.
- Extremely durable (B-120) Premium Fiberglass will not chip, fade, rust, shatter or peel for up to 25 years.
- Plastic signs (B-401) are ideal for use in light duty industrial, utility, commercial and institutional environments.
Specifications
ANSI Z535, OSHA 1910.145 | |
Surface Mount | |
DANGER Acetylene | |
Red on White |
1 Ea. | |
-40°F | |
Chemical and Hazardous Materials Sign |