Learn More
Brady™ Mild Sheet Steel Portable Metal Group Lock Box
Description
- Heavy steel lock box stores keys after machine or process is locked out
- Each authorized work team member adds a personal lock or tagout device on the group lock box
- Ensures that no one employee has access unless all employees have removed their locks or tags
- Accepts up to 13 individual locks
Specifications
Specifications
| Color | Red |
| Quantity | 1 Ea. |