missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brachyury Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5114917
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total Brachyury / Bry.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
Specifications
| Brachyury | |
| Polyclonal | |
| Unconjugated | |
| TBXT | |
| Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 20997, 20998, 292301, 6862 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| O15178, P20293 | |
| T, TBXT | |
| A synthesized peptide derived from human T(Accession O15178), corresponding to amino acid residues S321-L371. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction