missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brachyury Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA546984
This item is not returnable.
View return policy
Description
In direct ELISAs, approximately 25% cross-reactivity with recombinant human (rh) TBX-6 is observed, and approximately 5% cross-reactivity with rhTBX-2, rhTBX-5, and rhTBX-18 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
Specifications
| Brachyury | |
| Polyclonal | |
| Unconjugated | |
| TBXT | |
| Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 20997, 20998, 6862 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| ChIP Assay, Flow Cytometry, Immunohistochemistry (Frozen), Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 5% trehalose and No Preservative | |
| O15178, P20293 | |
| T, TBXT | |
| E. coli-derived recombinant human Brachyury Ser2-Glu202. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction