missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Brachyury Monoclonal Antibody (6C12)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA531920
This item is not returnable.
View return policy
Description
MA5-31920 has been tested in indirect ELISA.
The protein encoded by this gene is an embryonic nuclear transcription factor that binds to a specific DNA element, the palindromic T-site. It binds through a region in its N-terminus, called the T-box, and effects transcription of genes required for mesoderm formation and differentiation. The protein is localized to notochord-derived cells.
Specifications
| Brachyury | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 0.05% sodium azide | |
| O15178 | |
| TBXT | |
| Purified recombinant fragment of human T (AA: 218-352) expressed in E. coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Western Blot | |
| 6C12 | |
| Unconjugated | |
| TBXT | |
| Bra; brachyury; brachyury homolog; brachyury protein; brachyury-like 2; brachyury-like 3; cou; D17Mit170; Low; low ratio; Lr; me75; Protein T; RP11-459F1.1; SAVA; T; T brachyury homolog; T brachyury transcription factor; T, brachyury homolog; T1; T-box transcription factor T; TBXT; TFT; Tl2; Tl3 | |
| Mouse | |
| Protein G | |
| RUO | |
| 6862 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction