missing translation for 'onlineSavingsMsg'
Learn More

BQ 788, TRC

Product Code. 30331108
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 30331108

Brand: Toronto Research Chemicals TRCB6766705MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material sodium;(2R)-2-[[(2R)-2-[[(2S)-2-[[(2S,6R)-2,6-dimethylpiperidine-1-carbonyl]amino]-4,4-dimethyl-pentanoyl]amino]-3-(1-methoxycarbonylindol-3-yl)propanoyl]amino]hexanoate
CAS 156161-89-6
Molecular Formula C34 H50 N5 O7 . Na
Quantity 5 mg
Synonym BQ 788, cis-N-[N-[N-[(2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl]-1-(methoxycarbonyl)-D-tryptophyl]-D-norleucine Monosodium Salt, N-[[(2R,6S)-2,6-Dimethyl-1-piperidinyl]carbonyl]-4-methyl-L-leucyl-1-(methoxycarbonyl)-D-tryptophyl-D-norleucine Monosodium Salt, N-[(cis-2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl-1-(methoxycarbonyl)-D-tryptophyl-D-norleucine Monosodium Salt, (R)-2-((R)-2-((S)-2-((2S,6R)-2,6-Dimethylpiperidine-1-carboxamido)-4,4-dimethylpentanamido)-3-(1-(methoxycarbonyl)-1H-indol-3-yl)propanamido)hexanoic Acid Sodium Salt
InChI Formula InChI=1S/C34H51N5O7.Na/c1-8-9-16-25(31(42)43)35-29(40)26(18-23-20-38(33(45)46-7)28-17-11-10-15-24(23)28)36-30(41)27(19-34(4,5)6)37-32(44)39-21(2)13-12-14-22(39)3;/h10-11,15,17,20-22,25-27H,8-9,12-14,16,18-19H2,1-7H3,(H,35,40)(H,36,41)(H,37,44)(H,42,43);/q;+1/p-1/t21-,22+,25-,26-,27+;/m1./s1
SMILES [Na+].CCCC[C@@H](NC(=O)[C@@H](Cc1cn(C(=O)OC)c2ccccc12)NC(=O)[C@H](CC(C)(C)C)NC(=O)N3[C@H](C)CCC[C@@H]3C)C(=O)[O-]
IUPAC Name sodium;(2R)-2-[[(2R)-2-[[(2S)-2-[[(2S,6R)-2,6-dimethylpiperidine-1-carbonyl]amino]-4,4-dimethyl-pentanoyl]amino]-3-(1-methoxycarbonylindol-3-yl)propanoyl]amino]hexanoate
Molecular Weight (g/mol) 663.78
Formula Weight 663.3608
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.