missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BQ 788, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | sodium;(2R)-2-[[(2R)-2-[[(2S)-2-[[(2S,6R)-2,6-dimethylpiperidine-1-carbonyl]amino]-4,4-dimethyl-pentanoyl]amino]-3-(1-methoxycarbonylindol-3-yl)propanoyl]amino]hexanoate |
| CAS | 156161-89-6 |
| Molecular Formula | C34 H50 N5 O7 . Na |
| Quantity | 1 mg |
| Synonym | BQ 788, cis-N-[N-[N-[(2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl]-1-(methoxycarbonyl)-D-tryptophyl]-D-norleucine Monosodium Salt, N-[[(2R,6S)-2,6-Dimethyl-1-piperidinyl]carbonyl]-4-methyl-L-leucyl-1-(methoxycarbonyl)-D-tryptophyl-D-norleucine Monosodium Salt, N-[(cis-2,6-Dimethyl-1-piperidinyl)carbonyl]-4-methyl-L-leucyl-1-(methoxycarbonyl)-D-tryptophyl-D-norleucine Monosodium Salt, (R)-2-((R)-2-((S)-2-((2S,6R)-2,6-Dimethylpiperidine-1-carboxamido)-4,4-dimethylpentanamido)-3-(1-(methoxycarbonyl)-1H-indol-3-yl)propanamido)hexanoic Acid Sodium Salt |
| InChI Formula | InChI=1S/C34H51N5O7.Na/c1-8-9-16-25(31(42)43)35-29(40)26(18-23-20-38(33(45)46-7)28-17-11-10-15-24(23)28)36-30(41)27(19-34(4,5)6)37-32(44)39-21(2)13-12-14-22(39)3;/h10-11,15,17,20-22,25-27H,8-9,12-14,16,18-19H2,1-7H3,(H,35,40)(H,36,41)(H,37,44)(H,42,43);/q;+1/p-1/t21-,22+,25-,26-,27+;/m1./s1 |
| SMILES | [Na+].CCCC[C@@H](NC(=O)[C@@H](Cc1cn(C(=O)OC)c2ccccc12)NC(=O)[C@H](CC(C)(C)C)NC(=O)N3[C@H](C)CCC[C@@H]3C)C(=O)[O-] |
| IUPAC Name | sodium;(2R)-2-[[(2R)-2-[[(2S)-2-[[(2S,6R)-2,6-dimethylpiperidine-1-carbonyl]amino]-4,4-dimethyl-pentanoyl]amino]-3-(1-methoxycarbonylindol-3-yl)propanoyl]amino]hexanoate |
| Molecular Weight (g/mol) | 663.78 |
| Formula Weight | 663.3608 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction