missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BQ-788, MedChemExpress
Description
BQ-788 is a potent, selective ETB receptor antagonist with IC50 of 1.2 nM for inhibition of ET-1 binding to human Girardi heart cells, poorly inhibiting the binding to ETA receptors in human neuroblastoma cell line SK-N-MC cells with IC50 of 1300 nM.
Specifications
Specifications
| Chemical Name or Material | BQ-788 |
| CAS | 173326-37-9 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C34H51N5O7 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 170 mg/mL (264.88 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | CCCC[C@H](C(O)=O)NC([C@@H](CC1=CN(C(OC)=O)C2=CC=CC=C12)NC([C@H](CC(C)(C)C)NC(N3[C@H](C)CCC[C@@H]3C)=O)=O)=O |
| Molecular Weight (g/mol) | 641.8 |
| Formula Weight | 641.8 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?