missing translation for 'onlineSavingsMsg'
Learn More

Trajan™ BPX35 GC Capillary Column

Contains 35% Phenyl Polysilphenylene-siloxane. Trajan Scientific and Medical™ BPX35 Capillary GC Column is an MS premium, low bleed phase designed for trace analysis of herbicides and aromatics.

Brand:  Trajan™ 054724

Code : 13

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10075400

  • £530.00 / Each
Estimated Shipment: 14-06-2024
to see stock.
Add to basket

Description

Description

  • Mid-polarity column
  • Ideal for confirmational analysis
  • High temperature
  • Low bleed
  • Ideal for drugs of abuse, pharmaceuticals, PAHs, pesticides and herbicides
Specifications

Specifications

0.32 mm
SGE™ BPX 35
0.25 μm
0.25 μm
Mid polar
G42
Drugs of Abuse Analysis, Herbicides Analysis, Drugs of Abuse, Pharmaceuticals Analysis, Polynuclear Aromatic Hydrocarbons
10°C to 330/360°C
30 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers