missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trajan™ BPX Volatiles Column
SGE BPX Volatiles Column is a polar phase designed for EPA volatile organics analysis
Brand: Trajan™ 054980
Code : 13
Additional Details : Weight : 0.50000kg
Description
- Polar phase
- EPA organics analysis (EPA 624, 502.2, SW-846, 8240/8260)
- Ideal for environmental analyses, volatile organics, alcohol analysis
Specifications
0.25 mm | |
290/300°C | |
30 m |
BPX-Volatiles GC Column | |
1.4 μm |