missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPTES, MedChemExpress
Description
BPTES is an allosteric and selective glutaminase inhibitor with an IC50 of 0.16 μM.
Specifications
Specifications
| Chemical Name or Material | BPTES |
| Physical Form | Solid |
| CAS | 314045-39-1 |
| Color | White |
| Quantity | 10 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H24N6O2S3 |
| For Use With (Application) | Cancer-programmed cell death |
| Solubility Information | DMSO : 100 mg/mL (190.59 mM; Need ultrasonic) ∣H2O : ≥ 0.1 mg/mL (0.19 mM) |
| SMILES | O=C(NC1=NN=C(CCSCCC2=NN=C(NC(CC3=CC=CC=C3)=O)S2)S1)CC4=CC=CC=C4 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?