missing translation for 'onlineSavingsMsg'
Learn More

BPNT1 Antibody (3D4), Novus Biologicals™

Product Code. 18393729 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18393729 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18393729 Supplier Novus Biologicals Supplier No. H00010380M02

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BPNT1 Monoclonal antibody specifically detects BPNT1 in Human samples. It is validated for Western Blot, ELISA, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPNT1
Applications Western Blot, ELISA, Sandwich ELISA
Classification Monoclonal
Clone 3D4
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Sandwich ELISA
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_006076
Gene Alias 3'(2'), 5'-bisphosphate nucleotidase 1,3'(2')5'-bisphosphate nucleotidase 1, Bisphosphate 3'-nucleotidase 1, BPntase, EC 3.1.3, EC 3.1.3.7, PAP-inositol-1,4-phosphatase, PIP
Host Species Mouse
Immunogen BPNT1 (NP_006076, 1 a.a. ~ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MASSNTVLMRLVASAYSIAQKAGMIVRRVIAEGDLGIVEKTCATDLQTKADRLAQMSICSSLARKFPKLTIIGEEDLPSEEVDQELIEDSQWEEILKQPC
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Lipid and Metabolism
Primary or Secondary Primary
Gene ID (Entrez) 10380
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.