missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BPIL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100222
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BPIL1.
BPIL1 is a member of the lipid transfer/lipopolysaccharide binding protein (LT/LBP) family. This protein is highly expressed in hypertrophic tonsils.
Specifications
| BPIL1 | |
| Polyclonal | |
| Unconjugated | |
| BPIFB2 | |
| 2310034L21Rik; 2310069A01Rik; AV089125; Bactericidal/permeability-increasing protein-like 1; BPI fold containing family B member 2; BPI fold containing family B, member 2; BPI fold-containing family B member 2; Bpifb2; Bpil1; BPI-like 1; C20orf184; dJ726C3.2; Long palate, lung and nasal epithelium carcinoma-associated protein 2; LPLUNC2; RYSR; UNQ2489/PRO5776 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 80341 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8N4F0 | |
| BPIFB2 | |
| A synthesized peptide derived from human BPIFB2(Accession Q8N4F0), corresponding to amino acid residues L269-R319. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction