missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPIL1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92302-0.1ml
This item is not returnable.
View return policy
Description
BPIL1 Polyclonal antibody specifically detects BPIL1 in Mouse, Rat samples. It is validated for Western Blot
Specifications
| BPIL1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| bactericidal/permeability-increasing protein-like 1, C20orf184, Long palate, lung and nasal epithelium carcinoma-associated protein 2, LPLUNC2dJ726C3.2, RYSR | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 40-340 of human BPIFB2 (NP_079503.1). IGKAPLQRALQVTVPHFLDWSGEALQPTRIRILNVHVPRLHLKFIAGFGVRLLAAANFTFKVFRAPEPLELTLPVELLADTRVTQSSIRTPVVSISACSLFSGHANEFDGSNSTSHALLVLVQKHIKAVLSNKLCLSISNLVQGVNVHLGTLIGLNPVGPESQIRYSMVSVPTVTSDYISLEVNAVLFLLGKPIILPTDATPFVLPRHVGTEGSMATVGLSQQLFDSALLLLQKAGALNLDITGQLRSDDNLLNTSALGRLIPEVARQFPEPMPVVLKVRLGATPVAMLHTNNATLRLQPF | |
| 0.1 mL | |
| Cell Biology | |
| 80341 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction