missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPIFB1 Antibody (LPLUNC1/7059R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-14065JF635
This item is not returnable.
View return policy
Description
BPIFB1 Monoclonal antibody specifically detects BPIFB1 in Human samples. It is validated for ELISA,Western Blot,Immunohistochemistry (Paraffin)
Specifications
| BPIFB1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| bA49G10.6, chromosome 20 open reading frame 114, dJ1187J4.1, long palate, lung and nasal epithelium carcinoma associated 1, long palate, lung and nasal epithelium carcinoma-associated protein 1, LPLUNC1, MGC14597, VEMSGP, Von Ebner minor salivary gland protein | |
| Recombinant full-length human BPIFB1 protein (Uniprot: Q8TDL5) | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| ELISA, Western Blot, Immunohistochemistry (Paraffin) | |
| LPLUNC1/7059R | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| 92747 | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction