missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPI Antibody (971526) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB74681P
This item is not returnable.
View return policy
Description
BPI Monoclonal antibody specifically detects BPI in Human samples. It is validated for Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining)
Specifications
| BPI | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 671 | |
| Store at 4°C in the dark. | |
| IgG2b |
| Western Blot, Immunohistochemistry, Flow Cytometry (Intracellular Staining) | |
| 971526 | |
| Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry | |
| bactericidal permeability-increasing protein, bactericidal/permeability-increasing protein, CAP 57 | |
| Human embryonic kidney cell line HEK-293-derived transfected with human BPI, Val32-Lys487, Accession # P17213 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction