missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BPGM Antibody (2E11), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-59493DL594
This item is not returnable.
View return policy
Description
BPGM Monoclonal antibody specifically detects BPGM in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Specifications
| BPGM | |
| Monoclonal | |
| DyLight 594 | |
| 2,3-bisphosphoglycerate mutase, bisphosphoglycerate mutase, BPG-dependent PGAM, EC 3.1.3.132,3-bisphosphoglycerate synthase, EC 5.4.2.1, EC 5.4.2.4, erythrocyte 2,3-bisphosphoglycerate mutase2,3-bisphosphoglycerate mutase, erythrocyte | |
| Recombinant human BPGM (1-259aa) purified from E. coli. | |
| 0.1 mL | |
| Cancer, Cardiovascular Biology, Endocrinology, Signal Transduction | |
| 669 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| Western Blot, Flow Cytometry, ELISA, Immunofluorescence | |
| 200000000000 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction