missing translation for 'onlineSavingsMsg'
Learn More

BPDE Antibody (8E11), Novus Biologicals™

Product Code. 18371562 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18371562 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18371562 Supplier Novus Biologicals Supplier No. NBP311199

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BPDE Monoclonal specifically detects BPDE in Non-species specific samples. It is validated for Immunoassay.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BPDE
Applications Immunoassay
Classification Monoclonal
Clone 800000000000
Concentration 0.1 mg/ml
Conjugate Unconjugated
Dilution Immunoassay
Formulation 0.2 um filtered solution in PBS, 0.1% BSA
Gene Alias Benzo Pyrene Dihydrodiol Epoxide
Host Species Mouse
Immunogen The monoclonal antibody 8E11 recognizes free BPDE and DNA adducts.
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4°C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.