missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BP1 Antibody (OTI8A1), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70581PE
This item is not returnable.
View return policy
Description
BP1 Monoclonal antibody specifically detects BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry
Specifications
| BP1 | |
| Monoclonal | |
| PE | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry | |
| OTI8A1 | |
| Western Blot, Immunohistochemistry | |
| Beta protein 1, BP1distal-less homeo box 7, distal-less homeobox 4, DLX7distal-less homeo box 9, DLX8DLX9, homeobox protein DLX-4, Homeobox protein DLX-7, Homeobox protein DLX-8 | |
| Full length human recombinant protein of human DLX4(NP_612138) produced in E.coli. | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Growth and Development, Hematopoietic Stem Cell Markers, Immunology, Neuronal Cell Markers, Stem Cell Markers | |
| 1748 | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction